Learn More
Revvity Health Sciences Inc SCH 23390, [N-Methyl-3H]-, 250Ci (9.25MBq)
Supplier: Revvity Health Sciences Inc NET930250UC
SCH-23390 (halobenzazepine), tritiated on the N-methyl group. SCH-23390 acts as an antagonist at the dopamine D1 receptor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.