Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCH 39166 hydrobromide, Tocris Bioscience™
GSA_VA

Catalog No. p-4784522 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity D1-like antagonist

High affinity dopamine D1/D5 receptor antagonist; displays Ki values of 1.2, 2, 980, 5520, 80 and 731 nM for binding to D1, D5, D2, D4, 5-HT and α2a receptors, respectively.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1227675-51-5
Molecular Formula C19H21BrClNO
Molecular Weight (g/mol) 394.737
InChI Key GAUWIDFICGEZKR-UHFFFAOYSA-N
Synonym (6aS-trans)-11-Chloro-6,6a,7,8,9,13b-hexahydro-7-methyl-5H-benzo[d]naphth[2,1-b]azepin-12-ol hydrobromide
PubChem CID 73324728
IUPAC Name 11-chloro-7-methyl-5,6,6a,8,9,13b-hexahydronaphtho[1,2-a][3]benzazepin-12-ol;hydrobromide
SMILES CN1CCC2=CC(=C(C=C2C3C1CCC4=CC=CC=C34)O)Cl.Br

Specifications

Quantity 10 mg
Formula Weight 394.73
Percent Purity >99%
Chemical Name or Material SCH 39166 hydrobromide
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.