Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCH 50911, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective, competitive and orally active GABAB antagonist. Displays an IC50 of 1.1 μM at GABAB, approximately 60 times that of CGP 35348 (Cat. No. 1245) and no binding affinity for GABAA at concentrations up to 100 μM.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 173.21 |
Percent Purity | >98% |
Chemical Name or Material | SCH 50911 |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction