Learn More
TARGETMOL CHEMICALS INC SCH 58261 10MG
Supplier:  TARGETMOL CHEMICALS INC T6979
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH 58261 is a potent and selective A2a adenosine receptor antagonist.The Ki =2.3 nM for rat A2a and 2 nM is for bovine A2a. purity: 99%
Catalog No. 50-257-6397
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.