Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCH 58261, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
SCH 58261 is a potent and selective A2A adenosine receptor competitive antagonist (Ki = 1.3 nM). Displays 323-, 53- and 100-fold selectivity over A1, A2B and A3 receptors, respectively. Reduces melanoma tumor growth and TGF-β levels in WT and P2X7R-null mice.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 345.36 |
Percent Purity | >99% |
Chemical Name or Material | SCH 58261 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction