Learn More
Volu Sol Schaudinn's Fixative, Modified (Mercury Free), 16 oz / 500 mL, Volu-Sol™
Supplier: Volu Sol VXM016
Modified Schaudinn's Fixative, used as an on-slide parasite fixative for fresh fecal smears. Prevents distortion of parasitic elements. Contains no mercury, component of Volu-Sol's Parasitology Kit. Uses a copper sulfate formula
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.