Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SCNN1B Recombinant Protein

Catalog No. p-3721852 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg
25 μg

Human SCNN1B full-length ORF (AAH36352, 1 a.a. - 640 a.a.) recombinant protein with GST-tag at N-terminal

Nonvoltage-gated, amiloride-sensitive, sodium channels control fluid and electrolyte transport across epithelia in many organs. These channels are heteromeric complexes consisting of 3 subunits: alpha, beta, and gamma. This gene encodes the beta subunit.

  • Molecular weight: 96.14kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
  • Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue

Sequence:
MHVKKYLLKGLHRLQKGPGYTYKELLVWYCDNTNTHGPKRTICEGPKKKAMWFLLTLLFAALVCWQWGIFIRTYLSWEVSVSLSVGFKTMDFPAVTICNASPFKYSKIKHLLKDLDELMEAVLERILAPELSHANATRNLNFSIWNHTPLVLIDERNPHHPMVLDLFGDNHNGLTSSSASEKICNAHGCKMAMRLCSLNRTQCTFRNFTSATQALTEWYILQATNIFAQVPQQELVEMSYPGEQMILACLFGAEPCNYRNFTSIFYPHYGNCYIFNWGMTEKALPSANPGTEFGLKLILDIGQEDYVPFLASTAGVRLMLHEQRSYPFIRDEGIYAMSGTETSIGVLVDKLQRMGEPYSPCTVNGSEVPVQNFYSDYNTTYSIQACLRSCFQDHMIRNCNCGHYLYPLPRGEKYCNNRDFPDWAHCYSDLQMSVAQRETCIGMCKESCNDTQYKMTISMADWPSEASEDWIFHVLSQERDQSTNITLSRKGIVKLNIYFQEFNYRTIEESAANNIVWLLSNLGGQFGFWMGGSVLCLIEFGEIIIDFVWITIIKLVALAKSLRQRRAQASYAGPPPTVAELVEAHTNFGFQPDTAPRSPNTGPYPSEQALPIPGTPPPNYDSLRLQPLDVIESDSEGDAI

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH36352
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 6338
Molecular Weight (g/mol) 96.14
Name SCNN1B (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MHVKKYLLKGLHRLQKGPGYTYKELLVWYCDNTNTHGPKRTICEGPKKKAMWFLLTLLFAALVCWQWGIFIRTYLSWEVSVSLSVGFKTMDFPAVTICNASPFKYSKIKHLLKDLDELMEAVLERILAPELSHANATRNLNFSIWNHTPLVLIDERNPHHPMVLDLFGDNHNGLTSSSASEKICNAHGCKMAMRLCSLNRTQCTFRNFTSATQALTEWYILQATNIFAQVPQQELVEMSYPGEQMILACLFGAEPCNYRNFTSIFYPHYGNCYIFNWGMTEKALPSANPGTEFGLKLILDIGQEDYVPFLASTAGVRLMLHEQRSYPFIRDEGIYAMSGTETSIGVLVDKLQRMGEPYSPCTVNGSEVPVQNFYSDYNTTYSIQACLRSCFQDHMIRNCNCGHYLYPLPRGEKYCNNRDFPDWAHCYSDLQMSVAQRETCIGMCKESCNDTQYKMTISMADWPSEASEDWIFHVLSQERDQSTNITLSRKGIVKLNIYFQEFNYRTIEESAANNIVWLLSNLGGQFGFWMGGSVLCLIEFGEIIIDFVWITIIKLVALAKSLRQRRAQASYAGPPPTVAELVEAHTNFGFQPDTAPRSPNTGPYPSEQALPIPGTPPPNYDSLRLQPLDVIESDSEGDAI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ENaCb/ENaCbeta/SCNEB
Common Name SCNN1B
Gene Symbol SCNN1B
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.