Learn More
Scott Safety™ Pak-Link™ Programmer
Provides Scott Air-Pak™75™ and Air-Pak™ NXG7™ SCBA users the capability to wirelessly retrieve data that is actively being stored on their PASS devices (2000 most current events — per NFPA 1982, 2007).
Supplier: Scott Safety™ 7012471513
Description
- Retrieves PASS device stored data
- Improves firefighter safety and accountability
- Can change factory set I.D.’s on Pak-Tracker Firefighter Locator
- Comes complete with all hardware and software
- Easily connects to all computers with a USB port
Specifications
Pak-Link Programmer |