Learn More
Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek

Ideal for peak identification work
Supplier: Restek Corporation 34464
Description
- CGA 165 connection and can deliver large volumes of sample
- Scotty™ (Air Liquide) 48
- Contents: 48L
- Pressure: 300 psig (21 bar)
- Outlet Fitting: CGA 165
- Weight: 1.75 lb. (0.8kg)
- Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
- DOT Specifications: 39 NRC
All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements
Specifications
DOT 39 NRC | |
10.2 cm | |
100 ppm | |
Ideal for peak identification work | |
41 cm | |
1.75 lb. | |
0.8 kg |
Carbon Dioxide in Nitrogen | |
4 in. | |
48 L | |
16.25 in. | |
300 psi | |
12.68 gal. | |
Air Standard |
Safety and Handling
ShelfLife : 2 Years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.