Learn More
Glass Expansion Screws for Skimmer Interface TG1023-Ni or TG1023-Cu
Supplier:  Glass Expansion TG5002
Encompass_Preferred
Screws for Skimmer Interface 501907725 (3600813), or 501907724 (3601320). A packet of four screws compatible with Thermo X Series and X Series II skimmer cone interfaces. These should be replaced if they are corroded or worn.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2929
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.