Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyl 4-Hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. p-7130567 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 17696-61-6
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00059626
InChI Key ZUOTXZHOGPQFIU-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid sec-Butyl Ester, sec-Butylparaben
PubChem CID 86607
IUPAC Name butan-2-yl 4-hydroxybenzoate
SMILES CCC(C)OC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 60°C
Color White
Boiling Point 210°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material sec-Butyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.