Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sec-Butyl Alcohol
SCI_ED

Catalog No. p-4443125 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Solution

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Color Colorless
Density 0.808g/mL
Quantity 100 mL
Packaging Solution
Linear Formula CH3CHOHCH2CH3
Formula Weight 74.12
Chemical Name or Material Sec-Butyl Alcohol
ShelfLife 2 years
DOTInformation DOT Class 3, III, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.