Learn More
3M™ Secure Click™ Respirator P95 Particulate Cartridge and Hydrogen Fluoride with Nuisance Acid Gas Relief
Secure Click™ Hard Case D9000 Series Particulate Filters are approved for use with Secure Click™ Respirators in environments containing certain particles in the chemical manufacturing industry.
Supplier: 3M™ D3076HF
Description
- Approved for use with 3M™ Secure Click™ Respirators in environments containing certain particles
- Hard case recommended for harsh areas for easy decontamination and protection
- Hard case helps protect against sparks and high heat
- Filter case designed for certain spray applications
- May be used in a variety of industries such as aluminum processing, chemicals and oil and gas
- Common uses of D3076HF particulate cartridges include hydrogen fluoride and for filtering solids such as those from processing minerals, coal, iron ore, cotton, flour, and certain other substances
- May also be used for liquid or oil based particles from sprays that do not also emit harmful vapors and for metal fumes produced from welding, brazing, cutting and other operations involving heating of metals
- Recommended for relief from nuisance levels of acid gases such as sulfur dioxide and/or chlorine
Specifications
Particulate | |
Bayonet | |
Secure Click | |
Particulate Cartridge | |
95% | |
D3000 |
NIOSH Approved | |
Beige | |
P | |
No | |
Secure Click Half Facepiece HF-800 Reusable Respirators | |
100 Respirators/Pk. |