Learn More
Restek™ SeCure Y Connector Kits
SDP
Ferrules and Knurled Nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting even at temperatures as high as 400°C
$381.00 - $395.00
Specifications
Includes SeCure Y Connector Body (1), Knurled Nuts (3), Universal Y Press-Tight Union (1), and Ferrules (3)
Color Mixed
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
06-611-148
SDP
View Documents
Restek™
20276
0.18/0.25/0.28 mm ID Columns
Each for $395.00
Add to Cart
06-611-149
SDP
View Documents
Restek™
20278
0.45/0.53 mm ID Columns
Each for $381.00
Add to Cart
Description
Description
  • Combines the simplicity of a Y Press-Tight connector with the strength of a metal union
  • Connect two analytical columns to a transfer line or guard column
  • Use standard Y Press-Tight connectors and 1/16 in. graphite ferrules
  • Reliable seal integrity will not unexpectedly disconnect during temperature-programmed analyses
  • Open design allows visual confirmation of the seal for added confidence in the connection
  • Fits both Restek cage designs
Specifications
Specifications
SeCure Y Connector Body (1), Knurled Nuts (3), Universal Y Press-Tight Union (1), and Ferrules (3)
Mixed