Learn More
Electron Microscopy Sciences SEM Finder Grid SEMF1, Gold
Supplier:  Electron Microscopy Sciences 80101AU
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. Four quadrant markers tapered towards center. 100 radial sectors.
Material: Gold
Diameter: 10 mm
Thickness: ~50 µm
Catalog No. 50-287-66
missing translation for 'QuantityLeft'