Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 9-Division 25 mm Head with 9 mm Pin, Aluminum, 3.2 mm Pin diameter
Supplier: Electron Microscopy Sciences 7518340
9 divisions surface min stub mounts are for use with FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Mounts are made from lathe finished ultra-pure aluminum.
Head: 1"
Pin height: 9 mm
Pin diameter: 3.2 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.