Learn More
Electron Microscopy Sciences JEOL SEM Cylinder Mount 2" surface, 1/4" height, Aluminum
Supplier:  Electron Microscopy Sciences 75706
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Cylinder specimen mounts are for use with JEOL scanning electron microscopes. The mounts are made from ultra-pure aluminum with a standard lathe finish.
Head: 2"
Height: 1/4"
Catalog No. 50-192-7100