Learn More
Sempermed™ SemperCare™ Premium Latex Gloves
Shop All Sempermed USA Inc. Products- Examination grade
- Elasticity and comfortable fit
- Improved tactile sensitivity
- Polymer coated and easy to don
- Micro-rough textured surface
- Ideal for medical, dental, lab and vet applications
Specifications
Chemotherapeutic Rated
No
Applications
Healthcare, Laboratory
Color
Cream
Disposable
Single-use
Exterior Finish
Fully Textured
Finger Thickness (English)
4 mil
Finger Thickness (Metric)
0.10 mm
Laboratory/Cleanroom Use
No
Latex Free
No
Material
Latex
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.