Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sensidyne™ Detector Tubes: Ethylene
SDP

$316.85

Specifications

Components Measured Ethylene
Quantity 10
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 2
Catalog Number Mfr. No. Measurement Range Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Price Quantity & Availability  
18-069K
SDP
View Documents
Sensidyne™
108B
0.1 to 100ppm
Pack of 10 for $316.85
missing translation for 'QuantityLeft'
Add to Cart
 
18-069J
SDP
View Documents
Sensidyne™
108SA
20 to 1200ppm
Pack of 10 for $316.85
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Detector tubes are sold in packs of 10

Specifications

Specifications

Ethylene
10
32°F to 104°F
0°C to 40°C
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.