Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ ACCURUN™ HBV DNA Positive Control
Independent controls for HIV enhance the consistency and precision of test results, day-to-day and run-to-run
$998.00
Specifications
Format | Vial |
---|---|
Type | ACCURUN HBV DNA Positive Control |
Intended Use | Detect Hepatitis B Virus DNA |
Matrix | Human Serum or Plasma |
Catalog Number | Mfr. No. | Certifications/Compliance | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Quantity | Price | Quantity & Availability | ||||
22-554-523
|
SeraCare™
20200096 |
IVD, CE | 5 x 4 mL |
Pack of 5 for $998.00
|
|
||||
22-554-521
|
SeraCare™
20200095 |
RUO | 5 x 0.6 mL | N/A | N/A | N/A | |||
Description
- Human plasma based controls mimic patient samples
- Independent controls meet compliance requirements of regulatory agencies and professional organizations
- Liquid, ready-to-use: no reconstitution or special handling required
- Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
Specifications
Vial | |
ACCURUN HBV DNA Positive Control | |
Detect Hepatitis B Virus DNA | |
Human Serum or Plasma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title