Learn More
SeraCare™ ACCURUN™ Independent Positive Controls For Abbott ARCHITECT™
For use with in vitro diagnostic test methods that detect HIV-1 antigen
$855.00 - $1832.00
Specifications
Certifications/Compliance | ISO 9000 13485, IVD, CE |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Equipment) | Abbott Architect™ |
Type | ACCURUN 106 HIV-1 Antigen Positive Control |
Intended Use | Detect HIV-1 Antigen |
Alert:
Handle ACCURUN controls and all human blood products as though capable of transmitting infectious agents. Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN. Do not pipette by mouth; do not smoke, eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as though they contain infectious agents.
Catalog Number | Mfr. No. | Format | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Quantity | Price | Quantity & Availability | ||||
22-554-471
|
SeraCare™
20150065 |
Vial | 3 x 5 mL |
Pack of 3 for $855.00
|
|
||||
22-554-472
|
SeraCare™
20150066 |
Tube | 12 x 3.5 mL |
Pack of 12 for $1,832.00
|
|
||||
Description
ACCURUN 106 Series 1000 is prepared by diluting HIV-1 viral lysate derived from human cell culture in human serum or plasma nonreactive for HBsAg and negative for antibodies to HIV-1 and 2, HCV and HTLV
- For use with Abbott ARCHITECT
- For in vitro diagnostic use
- Helps to enhance quality control and quality assurance in testing labs
- Human plasma based controls mimic patient samples
- Independent controls meet compliance requirements of regulatory agencies and professional organizations
- Liquid, ready-to-use - no reconstitution or special handling required
- Extended shelf life means efficiency for labs, allowing assay performance to be monitored with same lot over an extended period of time
- All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
- Routine use of independent controls helps your lab monitor day-to-day test variability, lot-to-lot variability and staff performance
Specifications
ISO 9000 13485, IVD, CE | |
Abbott Architect™ | |
Detect HIV-1 Antigen | |
60 Days Open Vial |
2°C to 8°C | |
ACCURUN 106 HIV-1 Antigen Positive Control | |
Lysate derived from human cell culture in guman serum or plasma |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.