Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Series M Avantgarde.Line - Drying and Heating Chambers
SDP

With a maximum of +300°C and various programming options, the M series material test chambers are ideal for material and aging tests.

Supplier:  BINDER™ 90100373

 View more versions of this product

Catalog No. 01-672-878


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • Temperature range: +23°C above ambient temperature to +300°C
  • High temperature accuracy thanks to APT.line™ technology with increased heating output
  • Reinforced fan with adjustable speed
  • Electromechanical control of the exhaust air flap
  • Intuitive touchscreen controller with time-segment and real-time programming
  • 2 chrome-plated racks
  • Class 2 independent adjustable temperature safety device (DIN 12880) and class 3.1 independent temperature safety device (DIN 12880) with visual/acoustic alarm available for selection on the controller
  • Failsafe function – backup sensor Pt100
  • Computer interface: Ethernet
  • Internal data logger, measured values can be read out in open format via USB
Specifications

Specifications

9.04 cu. ft.
Series M Avantgarde.Line
M260-400V
32.05 x 33.07 x 37.87 in.
3.1 kW
M260-400V, Standard Series M Avantgarde.Line
88.18 lb.
25.59 x 19.88 x 30.71 in.
86 kg
9.6 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.