Learn More
ACROBiosystems Biotinylated domestic pig Serum Albumin Protein, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems DSAD82E1200UG
Encompass_Preferred
Biotinylated domestic pig Serum Albumin, His,Avitag (DSA-D82E1) is expressed from human 293 cells (HEK293). It contains AA Arg 19 - Ala 607 (Accession # A2THZ2-1).
Catalog No. 50-233-1674
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.