Learn More
Electron Microscopy Sciences Serum Bottles Amber Wheaton 500, 10 ml, 25 mm diameter, 54 mm height
Manufactured from Wheaton borosilicate molded glass, meets USP Type I standards.
Volume: 10 ml
Diameter: 25 mm
Height: 54 mm
Material: Wheaton "500"
Cap: 13 x 20 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.