Learn More
Electron Microscopy Sciences Serum Tube Vials Amber, 10 ml
Supplier: Electron Microscopy Sciences 6030210

Manufactured from Wheaton “320" borosilicate glass. Meets ASTM Type I, Class B and USP Type I Standards.
Volume: 10 ml
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.