Learn More
Crescent Chemical Co Inc SERVA IPG BLUESTRIP 3-10NL/7CM
Supplier: Crescent Chemical Co Inc SE43002.01
SERVA IPG BlueStrip 3-10 NL / 7 cm 70 x 3 x 0.5 mm SERVA IPG BlueStrips are dried gel strips with immobilized pH gradient used in high resolution 2D gel electrophoresis of proteins. The strips have to be rehydrated before use. The homogeneous polyacrylamide gel matrix is covalently
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.