Learn More
TARGETMOL CHEMICALS INC SESAMODIL 5MG
Supplier:  TARGETMOL CHEMICALS INC T28759
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Sesamodil (SD 3211) is a novel calcium antagonist that can be used to study hypertension. purity: 99%
Catalog No. 50-258-0495
missing translation for 'QuantityLeft'