Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEW 2871, Tocris Bioscience™

Catalog No. p-4784542 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-permeable, selective S1P1 receptor agonist

Novel, potent and selective sphingosine-1-phosphate 1 (S1P1) receptor agonist. Activates S1P1 receptor with an EC50 of 13 nM, but does not activate S1P2, S1P3, S1P4 or S1P5 receptors at concentrations up to 10 μM. Cell-permeable and active in vivo.

Chemical Identifiers

CAS 256414-75-2
Molecular Formula C20H10F6N2OS
Molecular Weight (g/mol) 440.36
MDL Number MFCD00096600
InChI Key OYMNPJXKQVTQTR-UHFFFAOYSA-N
Synonym 5-4-phenyl-5-trifluoromethyl-2-thienyl-3-3-trifluoromethyl phenyl-1,2,4-oxadiazole, s1p1 receptor agonist, sew2871, 5-4-phenyl-5-trifluoromethyl thiophen-2-yl-3-3-trifluoromethyl phenyl-1,2,4-oxadiazole, sphingosine-1-phosphate1 receptor s1p1 agonist, 5-4-phenyl-5-trifluoromethylthiophen-2-yl-3-3-trifluoromethylphenyl-1,2,4-oxadiazole, d0zu0o, sew hplc , solid
PubChem CID 4077460
IUPAC Name 5-[4-phenyl-5-(trifluoromethyl)thiophen-2-yl]-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
SMILES FC(F)(F)C1=C(C=C(S1)C1=NC(=NO1)C1=CC(=CC=C1)C(F)(F)F)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 440.36
Percent Purity >99%
Chemical Name or Material SEW 2871
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.