Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF 11, Tocris Bioscience™

Catalog No. p-4816703 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NPY Y2 receptor antagonist

SF 11 is an NPY Y2 antagonist (IC50 = 199 nM). Displays no affinity for Y1 receptor at concentrations up to 35 μM. Causes significant lethality in mice. Displays antidepressant effects in rats. Brain penetrant.

Chemical Identifiers

CAS 443292-81-7
Molecular Formula C27H30N2O2S
Molecular Weight (g/mol) 446.609
InChI Key PMEQBGAGFZDWQX-UHFFFAOYSA-N
Synonym n-4-ethoxyphenyl-4-hydroxy diphenyl methyl piperidine-1-carbothioamide, n-4-ethoxyphenyl-4-hydroxydiphenylmethyl-1-piperidinecarbothioamide, 4-hydroxy-diphenyl-methyl-piperidine-1-carbothioic acid 4-ethoxy-phenyl-amide, n-4-ethoxyphenyl-4-hydroxydiphenylmethyl piperidine-1-carbothioamide, regid_for_cid_2936384, sf hplc, 4-hydroxy diphenyl methyl-n-p-phenetyl-piperidine-1-carbothioamide, 1-4-ethoxyphenyl amino thioxomethyl 4-piperidyl diphenylmethan-1-ol, n-4-ethoxyphenyl-4-hydroxy diphenyl methyl-1-piperidinecarbothioamide, n-4-ethoxyphenyl-4-oxidanyl diphenyl methyl piperidine-1-carbothioamide
PubChem CID 2936384
IUPAC Name N-(4-ethoxyphenyl)-4-[hydroxy(diphenyl)methyl]piperidine-1-carbothioamide
SMILES CCOC1=CC=C(C=C1)NC(=S)N2CCC(CC2)C(C3=CC=CC=C3)(C4=CC=CC=C4)O

Specifications

Quantity 10 mg
Formula Weight 446.6
Percent Purity >99%
Chemical Name or Material SF 11
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.