Learn More
TARGETMOL CHEMICALS INC SFI003 5MG
Supplier:  TARGETMOL CHEMICALS INC T72068
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SFI003 is a novel SRSF3 inhibitor that exerts anticancer activity against colorectal cancer by modulating the SRSF3 / DHCR24 / ROS axis and driving apoptosis in CRC cells through the SRSF3 / DHCR24 / reactive oxygen species (ROS) axis. purity: 99%
Catalog No. 50-257-9248
missing translation for 'QuantityLeft'