Learn More
Trajan™ CTC PAL Removable Needle Syringe
Catalog No. SG006330 Shop All SGE Inc. Products
Click to view available options
Volume (Metric):
Equipped with PTFE-tipped plunger. For use with HPLC Autosamplers
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for High performance liquid chromatology (HPLC)
Specifications
Material
PTFE (Plunger)
Needle Gauge
22 G
Diameter (Metric) Inner Needle
0.37 mm
Diameter (Metric) Outer Needle
0.72 mm
Product Type
HPLC Autosampler Syringe
Volume (Metric)
250 μL
Needle Point Style
LC