Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ eVol™ XCHANGE Syringes

Catalog No. 03350322 Shop All SGE Inc. Products
Click to view available options
Volume (Metric):
Quantity:

XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.

  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available

Specifications

Needle Gauge

25 G

For Use With (Application)

Liquid handling procedure

Length (English)

1.9 in.

Length (Metric)

50 mm

Product Type

eVol™ Syringe

Quantity

3/Pk.

Volume (Metric)

100 μL

Length (Metric) Needle

50 mm

Needle Point Style

Bevel

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.