Learn More
MilliporeSigma™ Supelco™ SGE Needles for Luer Lock Syringes
Preferred for manual injection where piercing the septum in exactly the same place is difficult. The beveled tip is designed for optimum septum penetration and prevention of septum coring.
Supplier:  MilliporeSigma™ Supelco™ 26272
 View more versions of this product
Catalog No. 11-101-1223
Description
Description
Specifications
Specifications
Stainless Steel (Tip)
23 G
SGE Needles
5 Pk.
Bevel
NLL-7/23
0.63 mm
2
70 mm
Luer Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.