Learn More
MilliporeSigma™ Supelco™ SGE Syringes, Cone Tip
For use with Agilent/HP 7673, 7683 and 6850 ALS Autosamplers
Supplier:  MilliporeSigma™ Supelco™ 21543
 View more versions of this product
Catalog No. 11-101-0102
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Designed and tested to meet critical autosampler specifications
  • A unique plunger eliminates bending and seizing
  • Needle point style withstands multiple and fast septum injections
  • One of the most trouble free autosampler syringes available
Specifications
Specifications
23 G
Stainless Steel
1
10 μL
42 mm
Agilent/HP 7673, 7683 and 6850 ALS Autosampler
0.63 mm
10 F-HP-0.63
1 Ea.
SGE Syringe
Cone Tip
Fixed Needle (FN)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.