Learn More
Rockland Immunochemicals SHEEP BLOOD in Alsevers - R311-0050
SDP
Supplier:  Rockland Immunochemicals R3110050
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents.
Catalog No. 50-105-9133