Learn More
Eisco™ Shellbach Glass Burette with PTFE Stopcock, Sure Trace Certification
Schellbach Glass Burette, Class A with PTFE Stopcock
Supplier: Eisco™ CH0241LST
Description
- Accurate as per class A, DIN ISO 385, ASTM-E287 Compliant
- Unique features of the Schellbach design include a vertical white stripe with central blue ribbon to facilitate accurate reading of the meniscus and an intermediate PTFE stopcock which facilitates the complete drainage of excess liquid
- Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass
- Features chemical resistant blue printing
- All contents are included in a single package
Specifications
100 mL | |
Borosilicate Glass (3.3) | |
Buret |
86 cm | |
0.20 mL | |
Class A |