Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SID 7969543, Tocris Bioscience™

Catalog No. p-4816707 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor (IC50 values are 0.76, >33 and >33 μM at SF-1, RORα and VP16 respectively). Inhibits SF-1-dependent luciferase expression in HEK 293T cells in vitro (IC50 = 30 nM).

Chemical Identifiers

CAS 868224-64-0
Molecular Formula C24H24N2O7
Molecular Weight (g/mol) 452.463
InChI Key KWMBIIQCLUIHDI-UHFFFAOYSA-N
Synonym ethyl 2-2-2-2,3-dihydrobenzo b 1,4 dioxin-6-yl amino-2-oxoethyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2-2,3-dihydro-1,4-benzodioxin-6-yl amino-2-oxoethyl-1,2-dihydro-1-oxo-5-isoquinolinyl oxy propanoate, isoquinolinone scaffold, 2, d0aj5g, 2-2-2,3-dihydrobenzo 1,4 dioxin-6-ylcarbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yloxy propionic acid ethyl ester, sid hplc , solid, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-ylcarbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-yl carbamoyl methyl-1-oxo-1,2-dihydroisoquinolin-5-yl oxy propanoate, ethyl 2-2-2,3-dihydro-1,4-benzodioxin-6-yl carbamoyl methyl-1-oxoisoquinolin-5-yl oxy propanoate, ethyl 2-2-2-2,3-dihydro-benzo b 1,4 dioxine-6-yl amino-2-oxoethyl-1,2-dihydro-1-oxo-5-isoquinolinyl oxy propanoate
PubChem CID 4076092
IUPAC Name ethyl 2-[2-[2-(2,3-dihydro-1,4-benzodioxin-6-ylamino)-2-oxoethyl]-1-oxoisoquinolin-5-yl]oxypropanoate
SMILES CCOC(=O)C(C)OC1=CC=CC2=C1C=CN(C2=O)CC(=O)NC3=CC4=C(C=C3)OCCO4

Specifications

Quantity 10 mg
Formula Weight 452.46
Percent Purity >98%
Chemical Name or Material SID 7969543
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.