Learn More
TARGETMOL CHEMICALS INC SIGMA-LIGAND-1 5MG
Supplier:  TARGETMOL CHEMICALS INC T13510
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sigma-LIGAND-1 is a selective Sigma Receptor ligand with an IC50s of 16 nM 19 nM at the DTG site and the PPP site respectively. Sigma-LIGAND-1 has a Ki of 4000 nM at the dopamine D2 receptor. purity: 100%
Catalog No. 50-258-1341
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.