Learn More
Electron Microscopy Sciences Wet or Dry Silicone Carbide Disc 8", 240 Grit, Plain back
Supplier:  Electron Microscopy Sciences 5025005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 240 Grit
Backing: Plain
Catalog No. 50-997-146
missing translation for 'QuantityLeft'