Learn More
Electron Microscopy Sciences Wet or Dry Silicone Carbide Disc 8", 60 Grit, PSA backing
Supplier:  Electron Microscopy Sciences 5025501
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Pregrinding Wet/Dry Abrasive Papers (Discs) with premium Resin Bonding System for longer wear. Special adhesive eliminates need to transfer to plates. Uniform mineral size. Pull tabs for easy use.
Particles: 60 Grit
Backing: PSA
Catalog No. 50-997-152