Learn More
Chemglass Life Sciences Closure, Silicone Sponge, 13mm
SDP
Supplier:  Chemglass Life Sciences CLS1490013
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Closure, Silicone Sponge, 13mm. The silicone sponge closure is manufactured with a sealed in silicone sponge filter. Flange fits snugly over the necks of vessels permitting the exchange of gases, while reducing evaporation and preventing the passing of aerosols. Ideal for use with aerobic cultures. Reusable, autoclavable and will withstand dry heat sterilization up to 180°. Closure will withstand both dry heat sterilization and repeated autoclaving.CLS-1490-013 fits: 13-18mm Culture Tubes and 18mm Screw Necks
Catalog No. 50-121-5152
missing translation for 'QuantityLeft'