Learn More
Anton Paar 3mm ID x 5mm OD Silicone Tubing (1 meter)
Supplier:  Anton Paar 50814
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
This transparent silicone hose is used as a sample hose, for example, to lead the disposed sample to the waste vessel, or to connect the air pump outlet to the injection adapter. Please check the resistance of the hose against your samples and cleaning agents.Suitable for use with Anton Paar density meters DMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230
Catalog No. NC9651297
missing translation for 'QuantityLeft'