Learn More
Fisher Science Education™ Silver Nitrate Solution, 1.0N

$72.10 - $377.50
Chemical Identifiers
CAS | 7761-88-8, 7732-18-5 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID | 24470 |
ChEBI | CHEBI:32130 |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Description
This product is for educational use - no C of A is available.
Chemical Identifiers
7761-88-8, 7732-18-5 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Specifications
Colorless | |
< 2% w/v | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
Silver Nitrate Solution, 0.1N |
7761-88-8, 7732-18-5 | |
AgNO3 | |
1.0 to 1.1g/mL | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
Liquid |
Safety and Handling
ShelfLife : 2 years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.