Learn More
World Precision Instrument Teflon Coated Silver Wire-AGT10100
Supplier: World Precision Instrument AGT10100
- Teflon coated silver wire
- Available in 36, 30 and 26-27 AWG
- Available in precut lengths
- The Teflon coating is 150 micro-in. thick (4 μm). The Teflon coating is designed to reduce surface friction, only. It is not insulation.
- To get a quote:Â
- Call (866) 606-1974 to request a quote