Learn More
NEOGEN Simmons Citrate Agar, 500 g
SDP
Supplier:  NEOGEN 700003464
Encompass_Preferred
Simmons Citrate Agar is used for the differentiation of microorganisms on the basis of citrate utilization in a laboratory setting. Simmons Citrate Agar is not intended for use in the diagnosis of disease or other conditions in humans. Koser first developed a liquid medium for differentiating coliforms from fecal coliforms. Fecal coliforms were unable to use citrate as the sole source of carbon and inorganic ammonium salt as a sole source of nitrogen. Non-fecal coliforms, such as Enterobacter aerogenes could use citrate in such a medium with resultant alkalinity. Liquid medium had the disadvantage of appearing turbid when a large inoculum was used, although no growth had taken place. This observation led Simmons to devise a solid medium that eliminated the problem with turbidity. Simmons Citrate Agar is a modification of Koser’s medium, with the addition of bromothymol blue and 1.5% agar.For more information visit https://www.neogen.com/solutions/Microbiology/simmons-citrate-agar
Catalog No. 50-201-5639
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.