Learn More
Medchemexpress LLC HY-108335 5mg Medchemexpress, Sipatrigine CAS:130800-90-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1083355MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-108335 5mg Sipatrigine CAS:130800-90-7 Sipatrigine, a neuroprotective agent, is a glutamate release inhibitor, voltage-dependent sodium channel and calcium channel inhibitor, penetrating the central nervous system. Has potential to treat focal cerebral ischemia and stroke [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2316
missing translation for 'QuantityLeft'