Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SiteClick™ sDIBO Alkyne Kits for Antibody Labeling
SDP

Catalog No. p-7222418
Encompass
Click to view available options
Labeling Scale:
1 mg
100-250 μg
5 mg
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Biotin
Primary Amine
pHrodo Deep Red
pHrodo Red
Kit Contents:
Contains 1 vial of SiteClick pHrodo iFL Red sDIBO Alkyne for Antibody Labeling, 1 vial of DMSO, and 1 antibody concentrator

Easily attach Alexa Fluor™ pHrodo™ and Biotin sDIBO Alkyne tags to azido-modified antibodies using SiteClick™ chemistry.

Generate site-specific labeled antibody conjugates with SiteClick sDIBO Alkynes for Antibody Labeling. Our SiteClick sDIBO Antibody Labeling kits are optimized to be used in conjunction with the SiteClick Azido Modification kits (S20026, S10900, or S10901 sold separately) and contain sufficient reagents to perform a conjugation reaction of a SiteClick sDIBO Alkyne label using 100 µg, 1 mg, or 5 mg of azido-modified IgG antibody. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor dye, pHrodo dye, or biotin to an azido modified antibody. The label, when attached to the antibody, is located far away from its antigen binding domain.

Create your own covalently labeled antibody conjugates using the SiteClick sDIBO Alkyne labels, which are intended for use with azido-modified antibodies. SiteClick sDIBO Alkyne Antibody Labeling Kits include the sDIBO Alkyne and the antibody concentrator, enabling copper-free covalent attachment of Alexa Fluor and pHrodo dyes, biotin, or molecules of your choice. When used with the SiteClick Antibody Azido Modification Kit (available separately as Cat. No. S20026, S10900, S10901), labeling occurs on the heavy chain N-linked glycans, far away from the antigen-binding domain, enabling excellent reproducibility between separate labeling reactions and multiple antibody species and subtypes. The SiteClick sDIBO alkyne kits for antibody labeling are optimized for conjugation reactions, covalently linking 100 µg, 1 mg, or 5 mg of azide-modified IgG antibody.

The SiteClick Antibody Azido Modification Kit (Cat. No. S20026, S10900, S10901) is recommended for the attachment of an azide moiety to the heavy chains of an unlabeled IgG antibody. Other engineered antibodies containing azido moieties are also suitable.

When used in conjunction with the SiteClick Antibody Azido Modification Kit, the SiteClick sDIBO alkyne kits for antibody labeling give you the option of trying different fluorescent labels for your antibody, attaching other molecules via streptavidin, or attaching your own molecule via amine-reactive moieties, depending on your assay. Our sDIBO alkynes are improved versions of DIBO cyclooctynes, yielding conjugates that are less 'sticky' and emit lower signal background in biological samples.

Order Info

Store in freezer (-5 to -30°C) and protect from light.

Specifications

Kit Contents Contains 1 vial of SiteClick Alexa Fluor 488 sDIBO Alkyne for Antibody Labeling in 25 μL DMSO and 1 antibody concentrator
Filter Type GFP
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Chemical Reactivity Azide
Flow Cytometer Laser Lines Blue
Labeling Method Click Chemistry
Labeling Scale 100-250 μg
Shipping Condition Wet Ice
Reactive Moiety sDIBO Alkyne
Label or Dye Alexa Fluor 488
Color Green
Product Line Alexa Fluor™, SiteClick™
Labeling Target Azido modified antibodies
Excitation/Emission 490/525 nm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.