Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SKA 31
KCa3.1 (IK) and KCa2 (SK) channel activator
$163.10 - $674.50
Specifications
Purity | >99% |
---|---|
Product Type | SKA 31 |
Molecular Weight (g/mol) | 200.26 |
Inhibitors | SKA 31 |
Description
Activator of KCa3.1 and KCa2 channels (EC50 values are 260, 2900, 2900 nM for KCa3.1, KCa2.1 and KCa2.2 respectively). Potentiates acetylcholine-induced EDHF-type dilations of mouse carotid arteries and lowers blood pressure in normotensive and hypertensive mice.Specifications
>99% | |
SKA 31 | |
200.26 | |
SKA 31 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title