Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKF 81297 hydrobromide, Tocris Bioscience™
GSA_VA

Catalog No. p-4785574 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

D1 agonist

Dopamine D1-like receptor agonist. Centrally active following systemic administration in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 67287-39-2
Molecular Formula C16H17BrClNO2
Molecular Weight (g/mol) 370.671
InChI Key RMIJGBMRNYUZRG-UHFFFAOYSA-N
PubChem CID 11957706
IUPAC Name 9-chloro-5-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrobromide
SMILES C1CNCC(C2=CC(=C(C(=C21)Cl)O)O)C3=CC=CC=C3.Br

Specifications

Quantity 10 mg
Synonym skf 81297 hydrobromide, +/--6-chloro-pb hydrobromide, 6-chloro-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, y-6-chloro-pb hydrobromide, ?-6-chloro-pb hydrobromide, inverted question mark-6-chloro-pb hydrobromide, inverted question mark-skf-81297 hydrobromide, inverted exclamation marka-skf-81297 hydrobromide, inverted exclamation marka-6-chloro-pb hydrobromide, inverted exclamation marka-skf 81297 hydrobromide
Formula Weight 370.67
Percent Purity >98%
Chemical Name or Material SKF 81297 hydrobromide
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.