Learn More
TARGETMOL CHEMICALS INC SKF83959 5MG
Supplier: TARGETMOL CHEMICALS INC T60835
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKF83959 a benzazepine analog is a selective and potent partial agonist of the dopamine D1 receptor with Ki values of 1.18 7.56 920 and 399 nM for rat D1 D5 D2 and D3 receptors respectively.SKF83959 is a potent variant modulator of the sigma ((sigma))-1 receptor which ameliorates cognitive dysfunction for the study depression and Alzheimer's disease. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.